Assay Details
Target Gene Details
Entrez Gene ID: | 10478 |
Gene Name: | solute carrier family 25 member 17 |
Gene Aliases: |
PMP34 |
Location: |
Chr.22:40769630-40819399 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A17 | NM_001282726.1 | 7 | 747 | NP_001269655.1 |
| NM_001282727.1 | 4 | 475 | NP_001269656.1 | |
| NM_006358.3 | 6 | 694 | NP_006349.1 | |
| NR_104235.1 | 8 | 1006 | ||
| NR_104236.1 | 5 | 542 | ||
| NR_104237.1 | 5 | 584 | ||
| NR_104238.1 | 3 | 414 | ||
| AK094062.1 | 2 | 1278 | ||
| AK292924.1 | 6 | 632 | ||
| AK298215.1 | 4 | 363 | ||
| AK300553.1 | 7 | 657 | ||
| AU123445.1 | 5 | 501 | ||
| BC005957.1 | 6 | 632 | AAH05957.1 | |
| BC012998.2 | 6 | 614 | AAH12998.1 | |
| BQ440957.1 | 3 | 338 | ||
| BU595416.1 | ||||
| BX352732.2 | 5 | 397 | ||
| BX647991.1 | 5 | 474 | ||
| CR456577.1 | 6 | 610 | CAG30463.1 | |
| Y12860.1 | 6 | 664 | CAA73367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834205 | Chr.22:40593419 - 40777442 on Build GRCh38 | Gain+Loss |
|
| nsv834206 | Chr.22:40696722 - 40838881 on Build GRCh38 | Gain |
|
| nsv1062438 | Chr.22:40646389 - 40806134 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map