Assay Details
Target Gene Details
Entrez Gene ID: | 5306 |
Gene Name: | phosphatidylinositol transfer protein alpha |
Gene Aliases: |
HEL-S-36, PI-TPalpha, PITPN, VIB1A |
Location: |
Chr.17:1517989-1562816 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNA | NM_006224.3 | 12 | 1634 | NP_006215.1 |
| XM_011523939.1 | 11 | 1546 | XP_011522241.1 | |
| XM_011523940.1 | 12 | 2056 | XP_011522242.1 | |
| BC045108.1 | 12 | 1609 | AAH45108.1 | |
| BC068538.1 | 12 | 1517 | AAH68538.1 | |
| BC082976.1 | 12 | 1549 | AAH82976.1 | |
| D30036.1 | 12 | 1586 | BAA06276.1 | |
| EU794647.1 | 12 | 1609 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map