Assay Details
Target Gene Details
Entrez Gene ID: | 27434 |
Gene Name: | DNA polymerase mu |
Gene Aliases: |
Pol Mu, Tdt-N |
Location: |
Chr.7:44072062-44082540 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLM | NM_001284330.1 | 7 | 1089 | NP_001271259.1 |
| NM_001284331.1 | 8 | 1218 | NP_001271260.1 | |
| NM_013284.3 | 9 | 1329 | NP_037416.1 | |
| NR_104299.1 | 9 | 1300 | ||
| XM_005249711.2 | 6 | 880 | XP_005249768.1 | |
| XM_006715692.2 | 9 | 1300 | XP_006715755.1 | |
| XM_006715696.2 | 7 | 1030 | XP_006715759.1 | |
| XM_006715698.3 | 4 | 1184 | XP_006715761.1 | |
| XM_011515275.1 | 9 | 1315 | XP_011513577.1 | |
| XM_011515278.1 | 8 | 1165 | XP_011513580.1 | |
| XM_011515279.1 | 9 | 1315 | XP_011513581.1 | |
| XM_011515282.2 | 4 | 1181 | XP_011513584.1 | |
| XM_011515285.1 | 5 | 567 | XP_011513587.1 | |
| XM_017011998.1 | 8 | 1129 | XP_016867487.1 | |
| XM_017011999.1 | 7 | 1075 | XP_016867488.1 | |
| XM_017012000.1 | 8 | 1165 | XP_016867489.1 | |
| XM_017012001.1 | 7 | 1075 | XP_016867490.1 | |
| XM_017012002.1 | 8 | 1145 | XP_016867491.1 | |
| XM_017012003.1 | 6 | 893 | XP_016867492.1 | |
| AF176097.1 | 9 | 1269 | AAF26284.1 | |
| AJ131891.2 | 9 | 1272 | CAB65075.2 | |
| AK023002.1 | 9 | 1275 | BAB14353.1 | |
| AK092903.1 | 5 | 1052 | ||
| AK307634.1 | ||||
| BC026306.1 | 8 | 1872 | AAH26306.1 | |
| BC049202.1 | 7 | 1034 | ||
| BC062590.1 | 8 | 1142 | AAH62590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830983 | Chr.7:44068842 - 44201624 on Build GRCh38 | Loss |
|
| nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain |
|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv470210 | Chr.7:44045936 - 44139229 on Build GRCh38 | Loss |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map