Assay Details
Target Gene Details
Entrez Gene ID: | 84971 |
Gene Name: | autophagy related 4D cysteine peptidase |
Gene Aliases: |
APG4-D, APG4D, AUTL4 |
Location: |
Chr.19:10543894-10553423 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG4D | NM_001281504.1 | 9 | 1448 | NP_001268433.1 |
| NM_032885.5 | 9 | 1374 | NP_116274.3 | |
| NR_104024.1 | 9 | 1330 | ||
| NR_104025.1 | 8 | 1097 | ||
| XM_006722927.1 | 7 | 1029 | XP_006722990.1 | |
| AJ312332.1 | 9 | 1190 | CAC85951.1 | |
| AK056210.1 | 9 | 1349 | BAB71121.1 | |
| AK223383.1 | 9 | 1299 | BAD97103.1 | |
| AK294676.1 | 9 | 1448 | ||
| AK314712.1 | 9 | 1309 | ||
| BC007639.1 | 8 | 1020 | AAH07639.1 | |
| BC016845.1 | 4 | 332 | AAH16845.1 | |
| BC068992.1 | 9 | 1350 | AAH68992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map