Assay Details
Target Gene Details
Entrez Gene ID: | 102724663 |
Gene Name: | uncharacterized LOC102724663 |
Gene Aliases: |
- |
Location: |
Chr.12:81279189-81312422 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724663 | NR_120491.1 | |||
| BG186381.1 |
Target Gene Details
Entrez Gene ID: | 8499 |
Gene Name: | PTPRF interacting protein alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.12:81257975-81759748 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
More Information
Set Membership: |
|

Genomic Map