Assay Details
Target Gene Details
Entrez Gene ID: | 266727 |
Gene Name: | MAM domain containing glycosylphosphatidylinositol anchor 1 |
Gene Aliases: |
GPIM, MAMDC3 |
Location: |
Chr.6:37632508-37697990 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDGA1 | NM_153487.3 | 17 | 8655 | NP_705691.1 |
| XM_017010734.1 | 17 | 7617 | XP_016866223.1 | |
| AF478693.1 | 17 | 8655 | AAM77220.1 | |
| AK055376.1 | 1 | 2428 | ||
| AK090677.1 | 9 | 2624 | BAC03502.1 | |
| AK091650.1 | 3 | 2412 | ||
| AK092308.1 | 1 | 3099 | ||
| BX647180.1 | 5 | 5234 | ||
| BX647253.1 | 5 | 5234 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752124 | Chr.6:37448443 - 37712846 on Build GRCh38 | Gain |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map