Assay Details
Target Gene Details
Entrez Gene ID: | 80745 |
Gene Name: | THUMP domain containing 2 |
Gene Aliases: |
C2orf8 |
Location: |
Chr.2:39736060-39779276 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833870 | Chr.2:39609882 - 39798884 on Build GRCh38 | Gain |
|
| nsv833881 | Chr.2:39687418 - 39840116 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map