Assay Details
Target Gene Details
Entrez Gene ID: | 79856 |
Gene Name: | sorting nexin 22 |
Gene Aliases: |
- |
Location: |
Chr.15:64151698-64157481 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX22 | NM_024798.2 | NP_079074.2 | ||
| NR_073534.1 | ||||
| XM_005254677.3 | XP_005254734.1 | |||
| XM_017022581.1 | XP_016878070.1 | |||
| AK024014.1 | BAB14776.1 | |||
| AK126655.1 | BAC86630.1 | |||
| AL832838.1 | ||||
| AY044653.1 | AAK98767.1 | |||
| BC019655.1 | AAH19655.1 | |||
| BC030225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833036 | Chr.15:64072492 - 64215189 on Build GRCh38 | Loss |
|
| nsv827360 | Chr.15:64152078 - 64160104 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map