Assay Details
Target Gene Details
Entrez Gene ID: | 1191 |
Gene Name: | clusterin |
Gene Aliases: |
AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2 |
Location: |
Chr.8:27596917-27616717 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLU | NM_001831.3 | 9 | 2048 | NP_001822.3 |
| NR_038335.1 | 9 | 2183 | ||
| NR_045494.1 | 9 | 2042 | ||
| BM723283.1 | 1 | 61 | ||
| BQ181025.1 | 1 | 113 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831275 | Chr.8:27470773 - 27624088 on Build GRCh38 | Loss |
|
| esv3572652 | Chr.8:27589383 - 27606101 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map