Assay Details
Target Gene Details
Entrez Gene ID: | 23598 |
Gene Name: | POZ/BTB and AT hook containing zinc finger 1 |
Gene Aliases: |
MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23 |
Location: |
Chr.22:31325804-31346263 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PATZ1 | NM_014323.2 | 5 | 3191 | NP_055138.2 |
| NM_032050.1 | 4 | 3053 | NP_114439.1 | |
| NM_032052.1 | 5 | 3126 | NP_114441.1 | |
| AB208787.1 | 5 | 2423 | BAD92024.1 | |
| AF254083.1 | 4 | 3022 | AAG09032.1 | |
| AF254084.1 | 5 | 3095 | AAG09033.1 | |
| AF254085.1 | 5 | 3160 | AAG09034.1 | |
| AL096880.1 | 4 | 2387 | CAB51404.1 | |
| AY028384.1 | 4 | 2783 | AAK19024.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834176 | Chr.22:31286440 - 31468715 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map