Assay Details
Target Gene Details
Entrez Gene ID: | 6938 |
Gene Name: | transcription factor 12 |
Gene Aliases: |
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20 |
Location: |
Chr.15:56918090-57291261 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF12 | NM_001322151.1 | NP_001309080.1 | ||
| NM_001322152.1 | 1 | 53 | NP_001309081.1 | |
| NM_001322154.1 | 1 | 53 | NP_001309083.1 | |
| NM_001322156.1 | 1 | 53 | NP_001309085.1 | |
| NM_001322157.1 | 1 | 53 | NP_001309086.1 | |
| NM_001322158.1 | 1 | 53 | NP_001309087.1 | |
| NM_001322159.1 | 1 | 53 | NP_001309088.1 | |
| NM_003205.3 | 1 | 53 | NP_003196.1 | |
| NM_207036.1 | 1 | 53 | NP_996919.1 | |
| NM_207037.1 | 1 | 53 | NP_996920.1 | |
| NM_207038.1 | 1 | 53 | NP_996921.1 | |
| XM_011521959.2 | XP_011520261.1 | |||
| XM_011521960.2 | XP_011520262.1 | |||
| XM_011521961.2 | XP_011520263.1 | |||
| XM_011521962.2 | XP_011520264.1 | |||
| XM_011521963.2 | XP_011520265.1 | |||
| XM_017022520.1 | XP_016878009.1 | |||
| AK294617.1 | ||||
| AK302923.1 | ||||
| AK310708.1 | ||||
| AK312710.1 | ||||
| AL831981.2 | CAD89914.1 | |||
| BC050556.1 | AAH50556.1 | |||
| BU178185.1 | 1 | 64 | ||
| M80627.1 | AAA58632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569642 | Chr.15:56740239 - 56941457 on Build GRCh38 | Gain |
|
| nsv833020 | Chr.15:56763749 - 56923870 on Build GRCh38 | Gain+Loss |
|
| nsv457162 | Chr.15:56790808 - 56933568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map