Assay Details
Target Gene Details
Entrez Gene ID: | 139716 |
Gene Name: | GRB2 associated binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.X:154675249-154751583 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAB3 | NM_001081573.2 | 10 | 3641 | NP_001075042.1 |
| NM_001282283.1 | 9 | 3524 | NP_001269212.1 | |
| NM_080612.3 | 10 | 3638 | NP_542179.1 | |
| NR_104114.1 | 9 | 3130 | ||
| XM_005274648.1 | XP_005274705.1 | |||
| XM_006724804.1 | 11 | 2352 | XP_006724867.1 | |
| XM_011531103.1 | 11 | 3493 | XP_011529405.1 | |
| XM_011531105.1 | XP_011529407.1 | |||
| XM_011531106.1 | 10 | 3376 | XP_011529408.1 | |
| XM_017029276.1 | 11 | 2343 | XP_016884765.1 | |
| AK126283.1 | 9 | 3489 | ||
| AL832051.1 | 9 | 3089 | ||
| AY057989.1 | 10 | 3612 | AAL25825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv518228 | Chr.X:154601444 - 154719706 on Build GRCh38 | Gain |
|
| nsv521523 | Chr.X:154676120 - 154719706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map