Assay Details
Target Gene Details
Entrez Gene ID: | 2893 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 4 |
Gene Aliases: |
GLUR4, GLUR4C, GLURD, GluA4 |
Location: |
Chr.11:105609994-105982092 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIA4 | NM_000829.3 | 3 | 581 | NP_000820.3 |
| NM_001077243.2 | 3 | 581 | NP_001070711.2 | |
| NM_001077244.1 | 3 | 581 | NP_001070712.1 | |
| NM_001112812.1 | 2 | 449 | NP_001106283.1 | |
| NR_046356.1 | 2 | 449 | ||
| XM_005271518.3 | 3 | 625 | XP_005271575.1 | |
| XM_006718823.1 | 3 | 255 | XP_006718886.1 | |
| XM_011542775.2 | 3 | 625 | XP_011541077.1 | |
| XM_017017609.1 | 3 | 625 | XP_016873098.1 | |
| XM_017017610.1 | 3 | 625 | XP_016873099.1 | |
| AB209092.1 | 2 | 419 | BAD92329.1 | |
| BC045546.1 | 3 | 575 | AAH45546.1 | |
| BC114549.1 | 2 | 134 | AAI14550.1 | |
| BC142654.1 | 3 | 529 | ||
| BC146816.1 | 2 | 408 | ||
| BC150209.1 | 3 | 529 | ||
| DA391203.1 | 2 | 396 | ||
| DB473702.2 | 3 | 562 | ||
| U16129.1 | 2 | 135 | AAA95962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052674 | Chr.11:104111970 - 107043401 on Build GRCh38 | Loss |
|
| nsv472 | Chr.11:105587309 - 105616712 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map