Assay Details
Target Gene Details
Entrez Gene ID: | 8078 |
Gene Name: | ubiquitin specific peptidase 5 |
Gene Aliases: |
ISOT |
Location: |
Chr.12:6852118-6866632 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP5 | NM_001098536.1 | 20 | 2977 | NP_001092006.1 |
| NM_003481.2 | 20 | 2902 | NP_003472.2 | |
| AI289395.1 | 1 | 229 | ||
| BC004889.2 | 20 | 2881 | AAH04889.1 | |
| BC005139.2 | 20 | 2948 | AAH05139.1 | |
| U35116.1 | 20 | 2892 | AAA78934.1 | |
| U47927.1 | 20 | 2970 | AAC50465.1 | |
| X91349.1 | 20 | 2947 | CAA62690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss |
|
| nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss |
|
| esv3580405 | Chr.12:6826466 - 6903969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map