Assay Details
Target Gene Details
Entrez Gene ID: | 27343 |
Gene Name: | polymerase (DNA) lambda |
Gene Aliases: |
BETAN, POLKAPPA |
Location: |
Chr.10:101578882-101588270 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv964n100 | Chr.10:101363048 - 101579801 on Build GRCh38 | Gain |
|
| nsv1053934 | Chr.10:101409846 - 101691479 on Build GRCh38 | Gain |
|
| esv2750847 | Chr.10:101295225 - 101692888 on Build GRCh38 | Gain |
|
| esv3624382 | Chr.10:101510860 - 101643510 on Build GRCh38 | Gain |
|
| esv2760144 | Chr.10:101373136 - 101580586 on Build GRCh38 | Gain |
|
| nsv552056 | Chr.10:101441506 - 101584832 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map