Assay Details
Target Gene Details
Entrez Gene ID: | 50616 |
Gene Name: | interleukin 22 |
Gene Aliases: |
IL-21, IL-22, IL-D110, IL-TIF, ILTIF, TIFIL-23, TIFa, zcyto18 |
Location: |
Chr.12:68248245-68253501 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL22 | NM_020525.4 | NP_065386.1 | ||
| AF279437.1 | AAG22064.1 | |||
| AJ277247.1 | CAC06085.1 | |||
| AY358890.1 | AAQ89249.1 | |||
| BC066263.1 | AAH66263.1 | |||
| BC066264.1 | AAH66264.1 | |||
| BC066265.1 | AAH66265.1 | |||
| BC067509.1 | AAH67509.1 | |||
| BC067510.1 | AAH67510.1 | |||
| BC069112.1 | AAH69112.1 | |||
| BC069308.1 | AAH69308.1 | |||
| BC070261.1 | AAH70261.1 | |||
| CD559617.2 |
More Information
Set Membership: |
|

Genomic Map