Assay Details
Target Gene Details
Entrez Gene ID: | 1445 |
Gene Name: | c-src tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.15:74782064-74803198 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSK | NM_001127190.1 | NP_001120662.1 | ||
| NM_004383.2 | NP_004374.1 | |||
| XM_005254165.4 | XP_005254222.1 | |||
| XM_017021925.1 | XP_016877414.1 | |||
| AB451288.1 | ||||
| AB451420.1 | ||||
| AK127672.1 | ||||
| AK223626.1 | BAD97346.1 | |||
| AK290531.1 | ||||
| AK297694.1 | ||||
| AK298245.1 | ||||
| AK309344.1 | ||||
| AK312669.1 | ||||
| AY007162.1 | ||||
| BC104847.1 | AAI04848.1 | |||
| BC104875.1 | AAI04876.1 | |||
| BC106073.1 | AAI06074.1 | |||
| CR541960.1 | CAG46758.1 | |||
| X59932.1 | CAA42556.1 | |||
| X60114.1 | CAA42713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map