Assay Details
Target Gene Details
Entrez Gene ID: | 60484 |
Gene Name: | hyaluronan and proteoglycan link protein 2 |
Gene Aliases: |
BRAL1 |
Location: |
Chr.1:156601504-156626608 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAPLN2 | NM_021817.2 | NP_068589.1 | ||
| XM_005245415.2 | XP_005245472.1 | |||
| XM_011509853.2 | XP_011508155.1 | |||
| XM_017002020.1 | XP_016857509.1 | |||
| AB049054.1 | BAB17662.1 | |||
| AK090712.1 | ||||
| AK294131.1 | ||||
| BC029864.1 | AAH29864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|
| nsv521401 | Chr.1:156624864 - 156645402 on Build GRCh38 | Loss |
|
| nsv824864 | Chr.1:156624990 - 156625869 on Build GRCh38 | Gain |
|
| nsv824853 | Chr.1:156624803 - 156625289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map