Assay Details
Target Gene Details
Entrez Gene ID: | 60526 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
C2orf43, hLDAH |
Location: |
Chr.2:20682489-20823139 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDAH | NM_001282719.1 | 6 | 1205 | NP_001269648.1 |
| NM_001282720.1 | 6 | 1187 | NP_001269649.1 | |
| NM_001282721.1 | 6 | 1175 | NP_001269650.1 | |
| NM_001282722.1 | 5 | 1031 | NP_001269651.1 | |
| NM_001282723.1 | 6 | 1096 | NP_001269652.1 | |
| NM_001282724.1 | 5 | 970 | NP_001269653.1 | |
| NM_021925.3 | 7 | 1331 | NP_068744.1 | |
| NR_104233.1 | 5 | 940 | ||
| XM_011533014.2 | 7 | 1358 | XP_011531316.1 | |
| AK021672.1 | 3 | 553 | ||
| AK025473.1 | 7 | 1269 | BAB15142.1 | |
| AK300494.1 | 5 | 892 | ||
| AK301493.1 | ||||
| AK316204.1 | 5 | 1002 | ||
| AK316310.1 | ||||
| BC017473.1 | 7 | 1288 | AAH17473.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs111632024] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map