Assay Details
Target Gene Details
Entrez Gene ID: | 55526 |
Gene Name: | dehydrogenase E1 and transketolase domain containing 1 |
Gene Aliases: |
AMOXAD, CMT2Q |
Location: |
Chr.10:12068917-12123228 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHTKD1 | NM_018706.6 | NP_061176.3 | ||
| AK309810.1 | ||||
| BC002477.2 | AAH02477.1 | |||
| BC007955.2 | AAH07955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv674n100 | Chr.10:12077012 - 12134998 on Build GRCh38 | Gain |
|
| dgv37n21 | Chr.10:12024028 - 12123248 on Build GRCh38 | Gain |
|
| esv3622354 | Chr.10:12050805 - 12129371 on Build GRCh38 | Gain |
|
| nsv1035894 | Chr.10:12039222 - 12127501 on Build GRCh38 | Gain |
|
| dgv67n27 | Chr.10:12058736 - 12123248 on Build GRCh38 | Gain |
|
| nsv1035709 | Chr.10:12054410 - 12183299 on Build GRCh38 | Gain |
|
| dgv673n100 | Chr.10:12049505 - 12137652 on Build GRCh38 | Gain |
|
| nsv831785 | Chr.10:11928851 - 12124076 on Build GRCh38 | Gain |
|
| esv3891755 | Chr.10:12080356 - 12104859 on Build GRCh38 | Loss |
|
| nsv1046396 | Chr.10:12059681 - 12109949 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map