Assay Details
Target Gene Details
Entrez Gene ID: | 50808 |
Gene Name: | adenylate kinase 3 |
Gene Aliases: |
AK3L1, AK6, AKL3L, AKL3L1, FIX |
Location: |
Chr.9:4709556-4742043 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK3 | NM_001199852.1 | 4 | 2135 | NP_001186781.1 |
| NM_001199853.1 | 5 | 1954 | NP_001186782.1 | |
| NM_001199855.1 | 5 | 1961 | NP_001186784.1 | |
| NM_001199856.1 | 5 | 2027 | NP_001186785.1 | |
| NM_016282.3 | 5 | 2255 | NP_057366.2 | |
| AF100785.1 | 5 | 2033 | AAP97224.1 | |
| AK001553.1 | 5 | 2173 | BAA91753.1 | |
| AK027534.1 | 5 | 2047 | BAB55183.1 | |
| BX640794.1 | 1 | 70 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015376 | Chr.9:4686223 - 4737890 on Build GRCh38 | Gain |
|
| nsv972641 | Chr.9:4711171 - 4713505 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map