Assay Details
Target Gene Details
Entrez Gene ID: | 79709 |
Gene Name: | collagen beta(1-O)galactosyltransferase 1 |
Gene Aliases: |
GLT25D1 |
Location: |
Chr.19:17555594-17583162 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COLGALT1 | NM_024656.3 | 12 | 3224 | NP_078932.2 |
| XM_005260080.4 | 12 | 2870 | XP_005260137.1 | |
| XM_011528297.2 | 12 | 2862 | XP_011526599.1 | |
| XM_011528298.2 | 12 | 2963 | XP_011526600.1 | |
| XM_011528299.2 | 10 | 2737 | XP_011526601.1 | |
| AK025982.1 | 5 | 2002 | BAB15308.1 | |
| AK057140.1 | 2 | 2458 | ||
| AK075541.1 | 12 | 3116 | BAC11684.1 | |
| AL831905.1 | 1 | 938 | ||
| BC020492.1 | 4 | 1933 | AAH20492.1 | |
| BC108308.1 | 12 | 3179 | AAI08309.1 | |
| BU628916.1 | 1 | 501 |
More Information
Additional Information:
For this assay, SNP(s) [rs76660578] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map