Assay Details
Target Gene Details
Entrez Gene ID: | 10499 |
Gene Name: | nuclear receptor coactivator 2 |
Gene Aliases: |
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75 |
Location: |
Chr.8:70109762-70405390 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA2 | NM_001321703.1 | 17 | 3702 | NP_001308632.1 |
| NM_001321707.1 | 17 | 3548 | NP_001308636.1 | |
| NM_001321711.1 | 16 | 3423 | NP_001308640.1 | |
| NM_001321712.1 | 16 | 3340 | NP_001308641.1 | |
| NM_001321713.1 | 16 | 3443 | NP_001308642.1 | |
| NM_006540.3 | 17 | 3630 | NP_006531.1 | |
| XM_017012961.1 | 17 | 3617 | XP_016868450.1 | |
| XM_017012962.1 | 17 | 3641 | XP_016868451.1 | |
| XM_017012963.1 | 17 | 4854 | XP_016868452.1 | |
| XM_017012964.1 | 17 | 3909 | XP_016868453.1 | |
| XM_017012965.1 | 17 | 3574 | XP_016868454.1 | |
| XM_017012966.1 | 18 | 3838 | XP_016868455.1 | |
| XM_017012967.1 | 16 | 3552 | XP_016868456.1 | |
| XM_017012968.1 | 18 | 3639 | XP_016868457.1 | |
| XM_017012969.1 | 16 | 3670 | XP_016868458.1 | |
| XM_017012970.1 | 16 | 10089 | XP_016868459.1 | |
| XM_017012971.1 | 16 | 3801 | XP_016868460.1 | |
| XM_017012972.1 | 13 | 3132 | XP_016868461.1 | |
| AK298528.1 | 5 | 715 | ||
| BC114383.1 | 16 | 3497 | ||
| X97674.1 | 17 | 3575 | CAA66263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831352 | Chr.8:69999548 - 70182606 on Build GRCh38 | Gain |
|
| nsv6246 | Chr.8:70106516 - 70137048 on Build GRCh38 | Insertion |
|
| nsv6247 | Chr.8:70118114 - 70163017 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map