Assay Details
Target Gene Details
Entrez Gene ID: | 28982 |
Gene Name: | feline leukemia virus subgroup C cellular receptor 1 |
Gene Aliases: |
AXPC1, FLVCR, MFSD7B, PCA, PCARP |
Location: |
Chr.1:212858255-212899363 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLVCR1 | NM_014053.3 | 2 | 1009 | NP_054772.1 |
| XM_011509446.2 | 2 | 985 | XP_011507748.1 | |
| XM_011509447.2 | 2 | 985 | XP_011507749.1 | |
| XM_011509448.1 | 2 | 985 | XP_011507750.1 | |
| AF118637.1 | 2 | 1009 | AAD45243.1 | |
| AK001419.1 | 2 | 186 | BAA91679.1 | |
| AK314481.1 | 2 | 985 | ||
| BC048312.1 | 2 | 947 | AAH48312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv572n100 | Chr.1:212856472 - 212899671 on Build GRCh38 | Gain |
|
| dgv571n100 | Chr.1:212831823 - 212896221 on Build GRCh38 | Gain |
|
| nsv1008383 | Chr.1:212852204 - 212941552 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map