Assay Details
Target Gene Details
Entrez Gene ID: | 93183 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class M |
Gene Aliases: |
GPI-MT-I |
Location: |
Chr.1:160027672-160031993 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGM | NM_145167.2 | 1 | 3423 | NP_660150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587707 | Chr.1:160027538 - 160037887 on Build GRCh38 | Loss |
|
| esv2669935 | Chr.1:160027534 - 160037873 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map