Assay Details
Target Gene Details
Entrez Gene ID: | 199990 |
Gene Name: | Fanconi anemia core complex associated protein 20 |
Gene Aliases: |
C1orf86, FP7162 |
Location: |
Chr.1:2184460-2212720 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAP20 | NM_001146310.1 | 8 | 1515 | NP_001139782.1 |
| NM_001256945.1 | 3 | 645 | NP_001243874.1 | |
| NM_001282671.1 | 8 | 1596 | NP_001269600.1 | |
| NM_001282672.1 | 7 | 1465 | NP_001269601.1 | |
| NM_001282673.1 | 4 | 781 | NP_001269602.1 | |
| XM_017000554.1 | 4 | 756 | XP_016856043.1 | |
| XM_017000555.1 | XP_016856044.1 | |||
| XM_017000556.1 | XP_016856045.1 | |||
| AF469129.1 | 8 | 1562 | AAP97719.1 | |
| AK055593.1 | 7 | 1465 | BAB70965.1 | |
| AK127994.1 | 8 | 1515 | BAC87220.1 | |
| BG825658.1 | ||||
| BU192678.1 | 3 | 606 |
Target Gene Details
Entrez Gene ID: | 5590 |
Gene Name: | protein kinase C zeta |
Gene Aliases: |
PKC-ZETA, PKC2 |
Location: |
Chr.1:2050470-2185395 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCZ | NM_001033581.1 | 15 | 1948 | NP_001028753.1 |
| NM_001033582.1 | 15 | 1845 | NP_001028754.1 | |
| NM_001242874.1 | 15 | 1955 | NP_001229803.1 | |
| NM_002744.4 | 18 | 2160 | NP_002735.3 | |
| XM_017001790.1 | 18 | 2180 | XP_016857279.1 | |
| XM_017001791.1 | 15 | 1805 | XP_016857280.1 | |
| XM_017001796.1 | 15 | 1985 | XP_016857285.1 | |
| XM_017001798.1 | 15 | 1898 | XP_016857287.1 | |
| XM_017001799.1 | 15 | 2282 | XP_016857288.1 | |
| XM_017001801.1 | 13 | 1602 | XP_016857290.1 | |
| XM_017001804.1 | 15 | 2068 | XP_016857293.1 | |
| XM_017001805.1 | 15 | 2038 | XP_016857294.1 | |
| AK097627.1 | 13 | 1581 | ||
| AK131526.1 | 8 | 1695 | ||
| BC008058.2 | 18 | 2107 | AAH08058.1 | |
| BC014270.2 | 18 | 2160 | AAH14270.1 | |
| Z15108.1 | 18 | 1981 | CAA78813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
|
| dgv49n54 | Chr.1:2151050 - 2289783 on Build GRCh38 | Loss |
|
| nsv951530 | Chr.1:2153662 - 2229961 on Build GRCh38 | Deletion |
|
| nsv834557 | Chr.1:2117631 - 2311104 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map