Assay Details
Target Gene Details
Entrez Gene ID: | 10692 |
Gene Name: | retinal pigment epithelium-derived rhodopsin homolog |
Gene Aliases: |
- |
Location: |
Chr.4:109827994-109844707 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRH | NM_006583.2 | NP_006574.1 | ||
| AF012270.1 | AAC51757.1 | |||
| BC128120.1 | ||||
| BC128121.1 | ||||
| BC128401.1 |
More Information
Set Membership: |
|

Genomic Map