Assay Details
Target Gene Details
Entrez Gene ID: | 26512 |
Gene Name: | integrator complex subunit 6 |
Gene Aliases: |
DBI-1, DDX26, DDX26A, DICE1, HDB, INT6, Notchl2 |
Location: |
Chr.13:51353171-51453236 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS6 | NM_001039937.1 | 19 | 5192 | NP_001035026.1 |
| NM_001306091.1 | 20 | 5968 | NP_001293020.1 | |
| NM_012141.2 | 18 | 5684 | NP_036273.1 | |
| XM_011535040.2 | XP_011533342.1 | |||
| XM_011535041.1 | 17 | 5597 | XP_011533343.1 | |
| XM_017020540.1 | 17 | 5142 | XP_016876029.1 | |
| BE160101.1 | 1 | 87 | ||
| BG621629.1 | 1 | 476 | ||
| BU754174.1 | ||||
| FN136361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv832611 | Chr.13:51192336 - 51364178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map