Assay Details
Target Gene Details
Entrez Gene ID: | 10623 |
Gene Name: | RNA polymerase III subunit C |
Gene Aliases: |
RPC3, RPC62 |
Location: |
Chr.1:145823183-145844402 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3C | NM_001303456.1 | 12 | 1378 | NP_001290385.1 |
| NM_006468.7 | 12 | 1576 | NP_006459.3 | |
| XM_005272927.3 | 13 | 2349 | XP_005272984.1 | |
| XM_005272928.3 | 13 | 1265 | XP_005272985.1 | |
| AK223413.1 | 12 | 1339 | BAD97133.1 | |
| AY091463.1 | 11 | 1273 | AAM12033.1 | |
| BC002586.1 | 12 | 1425 | AAH02586.1 | |
| BC004424.1 | AAH04424.1 | |||
| U93867.1 | 12 | 1406 | AAB63675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015025 | Chr.1:145642988 - 145851919 on Build GRCh38 | Gain |
|
| nsv831381 | Chr.1:145784568 - 145960930 on Build GRCh38 | Gain |
|
| esv3587420 | Chr.1:145824369 - 145905171 on Build GRCh38 | Gain |
|
| esv3572008 | Chr.1:145837994 - 145846568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map