Assay Details
Target Gene Details
Entrez Gene ID: | 10845 |
Gene Name: | caseinolytic mitochondrial matrix peptidase chaperone subunit |
Gene Aliases: |
- |
Location: |
Chr.15:65148219-65185420 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLPX | NM_006660.4 | 10 | 1531 | NP_006651.2 |
| NR_133680.1 | 10 | 1485 | ||
| XM_011521164.2 | 9 | 1473 | XP_011519466.1 | |
| AJ006267.3 | 10 | 1336 | CAA06933.2 | |
| AK292316.1 | 10 | 1457 | ||
| AK308133.1 | 10 | 1451 | ||
| AL136922.1 | 10 | 1434 | CAB66856.1 | |
| BC130373.1 | 10 | 1327 | ||
| BC136487.1 | 10 | 1327 |
More Information
Set Membership: |
|

Genomic Map