Assay Details
Target Gene Details
Entrez Gene ID: | 25885 |
Gene Name: | RNA polymerase I subunit A |
Gene Aliases: |
A190, AFDCIN, RPA1, RPA194, RPO1-4, RPO14 |
Location: |
Chr.2:86026328-86106155 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1A | NM_015425.4 | 33 | 5326 | NP_056240.2 |
| AK025568.1 | 10 | 1505 | BAB15171.1 | |
| AK294447.1 | 22 | 3555 | ||
| AK302458.1 | 33 | 5051 | ||
| AK304131.1 | 20 | 3303 | ||
| AL117467.1 | 2 | 73 | CAB55942.1 | |
| BC017115.1 | 10 | 1511 | ||
| BC117173.1 | 33 | 4979 | ||
| BC126303.1 | 33 | 4979 | ||
| BC143345.1 | 33 | 4979 | ||
| U33460.1 | 33 | 5008 | AAC99959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751903 | Chr.2:86026419 - 86278330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map