Assay Details
Target Gene Details
Entrez Gene ID: | 1411 |
Gene Name: | crystallin beta A1 |
Gene Aliases: |
CRYB1, CTRCT10 |
Location: |
Chr.17:29246857-29254494 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYBA1 | NM_005208.4 | NP_005199.2 | ||
| XM_017024198.1 | XP_016879687.1 | |||
| BC069537.1 | AAH69537.1 | |||
| BC113693.1 | ||||
| BU735083.1 | ||||
| KM230602.1 | ||||
| KM230603.1 | ||||
| KM230604.1 | ||||
| KM230605.1 | ||||
| KR336660.1 | ||||
| KR336661.1 | ||||
| U59058.1 | AAC50971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953873 | Chr.17:29250583 - 29259382 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map