Assay Details
Target Gene Details
Entrez Gene ID: | 5438 |
Gene Name: | RNA polymerase II subunit I |
Gene Aliases: |
RPB9, hRPB14.5 |
Location: |
Chr.19:36113709-36115304 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2I | NM_006233.4 | NP_006224.1 | ||
| AK312206.1 | ||||
| BC017112.1 | AAH17112.1 | |||
| BC067794.1 | AAH67794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833815 | Chr.19:36028901 - 36134034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map