Assay Details
Target Gene Details
Entrez Gene ID: | 5610 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 2 |
Gene Aliases: |
EIF2AK1, PKR, PPP1R83, PRKR |
Location: |
Chr.2:37099210-37157065 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2AK2 | NM_001135651.2 | 13 | 1443 | NP_001129123.1 |
| NM_001135652.2 | 10 | 1013 | NP_001129124.1 | |
| NM_002759.3 | 13 | 1677 | NP_002750.1 | |
| XM_011532987.2 | 13 | 1377 | XP_011531289.1 | |
| XM_017004503.1 | 13 | 1373 | XP_016859992.1 | |
| AF086566.1 | 1 | 48 | ||
| AK290655.1 | 13 | 1363 | ||
| AK313818.1 | 12 | 1200 | ||
| AY302136.1 | 10 | 997 | AAP57628.1 | |
| BC007769.2 | AAH07769.1 | |||
| BC022314.1 | 12 | 1172 | ||
| BC040851.1 | 13 | 1443 | AAH40851.1 | |
| BC057805.1 | 13 | 1348 | AAH57805.1 | |
| BC093676.1 | 11 | 1148 | AAH93676.1 | |
| BC101475.1 | 11 | 1148 | AAI01476.1 | |
| BC143230.1 | 11 | 1148 | ||
| M35663.1 | 12 | 1306 | AAA36409.1 | |
| M85294.1 | 11 | 1150 | AAA18253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511779 | Chr.2:37119775 - 37121900 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map