Assay Details
Target Gene Details
Entrez Gene ID: | 84701 |
Gene Name: | cytochrome c oxidase subunit 4I2 |
Gene Aliases: |
COX4, COX4-2, COX4B, COX4L2, COXIV-2, dJ857M17.2 |
Location: |
Chr.20:31637856-31645009 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX4I2 | NM_032609.2 | NP_115998.2 | ||
| XM_005260579.4 | XP_005260636.1 | |||
| XM_005260580.4 | XP_005260637.1 | |||
| XM_005260581.3 | XP_005260638.1 | |||
| AF257180.1 | AAK49333.1 | |||
| BC057779.1 | AAH57779.1 | |||
| HE647859.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470546 | Chr.20:31245806 - 31647499 on Build GRCh38 | Gain |
|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
| esv2763228 | Chr.20:31247246 - 31651719 on Build GRCh38 | Gain |
|
| nsv585911 | Chr.20:31305698 - 31688103 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map