Assay Details
Target Gene Details
Entrez Gene ID: | 4209 |
Gene Name: | myocyte enhancer factor 2D |
Gene Aliases: |
- |
Location: |
Chr.1:156463721-156500842 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2D | NM_001271629.1 | 11 | 4182 | NP_001258558.1 |
| NM_005920.3 | 12 | 4484 | NP_005911.1 | |
| XM_005245169.4 | 12 | 4466 | XP_005245226.1 | |
| XM_005245170.3 | 12 | 4491 | XP_005245227.1 | |
| XM_006711330.3 | 12 | 4195 | XP_006711393.1 | |
| XM_006711332.3 | 12 | 4479 | XP_006711395.1 | |
| XM_006711333.2 | 11 | 4458 | XP_006711396.1 | |
| XM_006711334.3 | 11 | 4456 | XP_006711397.1 | |
| XM_011509569.2 | 12 | 4516 | XP_011507871.1 | |
| XM_017001314.1 | 11 | 4600 | XP_016856803.1 | |
| XM_017001315.1 | 11 | 4195 | XP_016856804.1 | |
| AB210005.1 | 11 | 4734 | BAE06087.1 | |
| AK027180.1 | 1 | 437 | ||
| BC054520.1 | 12 | 4379 | AAH54520.1 | |
| BX640792.1 | 1 | 925 | ||
| BX647226.1 | 1 | 2221 |
More Information
Set Membership: |
|

Genomic Map