Assay Details
Target Gene Details
Entrez Gene ID: | 161198 |
Gene Name: | C-type lectin domain family 14 member A |
Gene Aliases: |
C14orf27, CEG1, EGFR-5 |
Location: |
Chr.14:38254000-38256370 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC14A | NM_175060.2 | NP_778230.1 | ||
| AY358395.1 | AAQ88761.1 | |||
| BX248017.1 | CAD62342.1 | |||
| BX423057.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564399 | Chr.14:38222364 - 38283547 on Build GRCh38 | Gain |
|
| nsv832773 | Chr.14:38143573 - 38315799 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map