Assay Details
Target Gene Details
Entrez Gene ID: | 51603 |
Gene Name: | methyltransferase like 13 |
Gene Aliases: |
5630401D24Rik, CGI-01, KIAA0859, feat |
Location: |
Chr.1:171781504-171823962 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL13 | NM_001007239.1 | 3 | 884 | NP_001007240.1 |
| NM_014955.2 | 3 | 1027 | NP_055770.1 | |
| NM_015935.4 | 3 | 1352 | NP_057019.3 | |
| XM_017001455.1 | 2 | 846 | XP_016856944.1 | |
| XM_017001456.1 | 2 | 846 | XP_016856945.1 | |
| XM_017001457.1 | 2 | 846 | XP_016856946.1 | |
| XM_017001458.1 | 3 | 1469 | XP_016856947.1 | |
| XM_017001459.1 | 3 | 1019 | XP_016856948.1 | |
| AB020666.2 | 3 | 1107 | ||
| AB242174.1 | 3 | 1277 | ||
| AB242175.1 | 3 | 1265 | ||
| AF132936.1 | 3 | 1277 | AAD27711.1 | |
| AK001172.1 | 1 | 39 | ||
| AK027552.1 | 3 | 1271 | ||
| AK027621.1 | 3 | 966 | BAB55240.1 | |
| AK027886.1 | 2 | 823 | ||
| AK074552.1 | BAC11055.1 | |||
| AK129561.1 | 2 | 700 | ||
| AK223493.1 | 3 | 1313 | BAD97213.1 | |
| AK291740.1 | 3 | 1299 | ||
| AK304471.1 | 4 | 1133 | ||
| AL049669.1 | 3 | 809 | CAB41243.1 | |
| BC029083.1 | 3 | 1106 | AAH29083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518222 | Chr.1:171783899 - 171794409 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map