Assay Details
Target Gene Details
Entrez Gene ID: | 80162 |
Gene Name: | protein-glucosylgalactosylhydroxylysine glucosidase |
Gene Aliases: |
ATHL1 |
Location: |
Chr.11:287760-295688 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGGHG | NM_025092.4 | NP_079368.3 | ||
| XM_011520383.2 | XP_011518685.1 | |||
| XM_011520384.2 | XP_011518686.1 | |||
| XM_011520385.2 | XP_011518687.1 | |||
| XM_011520386.2 | XP_011518688.1 | |||
| XM_011520387.2 | XP_011518689.1 | |||
| XM_017018355.1 | XP_016873844.1 | |||
| AK026288.1 | BAB15431.1 | |||
| AK074155.1 | BAB84981.1 | |||
| AK090428.1 | BAC03409.1 | |||
| AK296137.1 | ||||
| BC109257.2 | AAI09258.1 | |||
| BC109258.2 | AAI09259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891905 | Chr.11:269147 - 336332 on Build GRCh38 | Gain |
|
| nsv516809 | Chr.11:290233 - 295876 on Build GRCh38 | Loss |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv1012n100 | Chr.11:198510 - 427836 on Build GRCh38 | Gain |
|
| esv989344 | Chr.11:291680 - 292918 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map