Assay Details
Target Gene Details
Entrez Gene ID: | 5801 |
Gene Name: | protein tyrosine phosphatase, receptor type R |
Gene Aliases: |
EC-PTP, PCPTP1, PTP-SL, PTPBR7, PTPRQ |
Location: |
Chr.12:70638073-70920804 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPRR | NM_001207015.1 | 13 | 1704 | NP_001193944.1 |
NM_001207016.1 | 11 | 1494 | NP_001193945.1 | |
NM_002849.3 | 14 | 2360 | NP_002840.2 | |
NM_130846.2 | 10 | 1626 | NP_570897.2 | |
NR_073474.1 | 11 | 1850 | ||
XM_011538615.2 | 14 | 1956 | XP_011536917.1 | |
AK091647.1 | 5 | 599 | ||
AK295951.1 | 13 | 1694 | ||
AK304457.1 | 11 | 1453 | ||
AK312376.1 | ||||
BC110900.1 | 10 | 1323 | AAI10901.1 | |
BC119777.1 | 11 | 1302 | ||
BC122866.1 | 11 | 1302 | ||
BX571751.1 | 10 | 1679 | CAE11876.1 | |
CR749836.1 | 5 | 759 | CAH18692.1 | |
D64053.1 | 14 | 2362 | BAA10930.1 | |
U42361.1 | 14 | 2120 | AAD09447.1 | |
U77916.1 | AAB54006.1 | |||
U77917.1 | 10 | 1603 | AAB54007.1 | |
X82635.1 | 10 | 1584 | CAA57957.1 | |
Z79693.1 | 14 | 2031 | CAB01957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32743 | Chr.12:70481471 - 70719432 on Build GRCh38 | Gain |
![]() ![]() |
nsv509472 | Chr.12:70606856 - 70653543 on Build GRCh38 | Insertion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
