Assay Details
Target Gene Details
Entrez Gene ID: | 84109 |
Gene Name: | pyroglutamylated RFamide peptide receptor |
Gene Aliases: |
AQ27, GPR103, SP9155 |
Location: |
Chr.4:121328642-121381029 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRFPR | NM_198179.2 | 6 | 1588 | NP_937822.2 |
| AB109629.1 | 6 | 1209 | BAC98938.1 | |
| BC128133.1 | 6 | 1209 | ||
| JF810892.1 | 6 | 1209 |
More Information
Set Membership: |
|

Genomic Map