Assay Details
Target Gene Details
Entrez Gene ID: | 148327 |
Gene Name: | cAMP responsive element binding protein 3 like 4 |
Gene Aliases: |
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL |
Location: |
Chr.1:153967679-153974364 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L4 | NM_001255978.1 | 1 | 230 | NP_001242907.1 |
| NM_001255980.1 | 1 | 230 | NP_001242909.1 | |
| NM_130898.3 | 1 | 230 | NP_570968.1 | |
| NR_045658.1 | 1 | 230 | ||
| XM_006711172.2 | 1 | 178 | XP_006711235.1 | |
| XM_017000372.1 | XP_016855861.1 | |||
| AB052778.1 | 1 | 148 | BAC45035.1 | |
| AK297703.1 | 1 | 199 | ||
| AK304665.1 | 1 | 155 | ||
| AK309767.1 | 1 | 161 | ||
| AY049977.1 | 1 | 166 | AAL13157.1 | |
| BC038962.1 | 1 | 152 | AAH38962.1 | |
| DB012316.1 | 1 | 230 |
Target Gene Details
Entrez Gene ID: | 27173 |
Gene Name: | solute carrier family 39 member 1 |
Gene Aliases: |
ZIP1, ZIRTL |
Location: |
Chr.1:153959099-153968184 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A1 | NM_001271960.1 | NP_001258889.1 | ||
| AF151829.1 | AAD34066.1 | |||
| DB001496.1 |
More Information
Set Membership: |
|

Genomic Map