Assay Details
Target Gene Details
Entrez Gene ID: | 8507 |
Gene Name: | ectodermal-neural cortex 1 |
Gene Aliases: |
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10 |
Location: |
Chr.5:74627406-74641424 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENC1 | NM_001256574.1 | 4 | 3510 | NP_001243503.1 |
| NM_001256575.1 | 4 | 3498 | NP_001243504.1 | |
| NM_001256576.1 | 3 | 3237 | NP_001243505.1 | |
| NM_003633.3 | 3 | 3373 | NP_003624.1 | |
| NR_046318.1 | 2 | 1558 | ||
| XM_011543696.2 | 4 | 3581 | XP_011541998.1 | |
| XM_011543697.2 | 4 | 3427 | XP_011541999.1 | |
| AF059611.1 | 3 | 2641 | AAC26109.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830351 | Chr.5:74485729 - 74637007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map