Assay Details
Target Gene Details
Entrez Gene ID: | 9064 |
Gene Name: | mitogen-activated protein kinase kinase kinase 6 |
Gene Aliases: |
ASK2, MAPKKK6, MEKK6 |
Location: |
Chr.1:27355179-27368037 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K6 | NM_001297609.1 | 16 | 2504 | NP_001284538.1 |
| NM_004672.4 | 17 | 2528 | NP_004663.3 | |
| XM_017002771.1 | 15 | 1909 | XP_016858260.1 | |
| XM_017002772.1 | 17 | 3699 | XP_016858261.1 | |
| AB167411.1 | 17 | 2259 | BAD12485.1 | |
| AB208805.1 | 16 | 1973 | BAD92042.1 | |
| AF100318.1 | 14 | 1673 | AAD05304.1 | |
| BC015914.1 | 13 | 1430 | AAH15914.1 | |
| BC109032.2 | 8 | 807 | AAI09033.1 | |
| BC129950.1 | 9 | 958 | ||
| BC129951.1 | 9 | 958 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map