Assay Details
Target Gene Details
Entrez Gene ID: | 5581 |
Gene Name: | protein kinase C epsilon |
Gene Aliases: |
PKCE, nPKC-epsilon |
Location: |
Chr.2:45651315-46187990 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCE | NM_005400.2 | 1 | 240 | NP_005391.1 |
| XM_005264428.1 | 2 | 269 | XP_005264485.1 | |
| XM_005264431.3 | 1 | 829 | XP_005264488.1 | |
| XM_017004492.1 | 1 | 829 | XP_016859981.1 | |
| AK313842.1 | 1 | 139 | ||
| AL832345.1 | 1 | 370 | ||
| BC109033.2 | 1 | 178 | AAI09034.1 | |
| BC109034.2 | 1 | 178 | AAI09035.1 | |
| X65293.1 | 1 | 58 | CAA46388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3804n100 | Chr.2:45180603 - 45750333 on Build GRCh38 | Gain |
|
| esv2751894 | Chr.2:45141551 - 45772680 on Build GRCh38 | Gain |
|
| dgv1119e212 | Chr.2:45181796 - 45749281 on Build GRCh38 | Gain |
|
| nsv1013787 | Chr.2:45541783 - 45738705 on Build GRCh38 | Gain |
|
| esv2762545 | Chr.2:45419685 - 46137453 on Build GRCh38 | Gain |
|
| nsv581741 | Chr.2:45622938 - 45681498 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map