Assay Details
Target Gene Details
Entrez Gene ID: | 51105 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, TDRD20B, URLC1 |
Location: |
Chr.8:132775358-132848807 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831463 | Chr.8:132723206 - 132905755 on Build GRCh38 | Gain |
|
| nsv1016348 | Chr.8:132769270 - 132803626 on Build GRCh38 | Loss |
|
| dgv7318n100 | Chr.8:132769940 - 132860065 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map