Assay Details
Target Gene Details
Entrez Gene ID: | 1191 |
Gene Name: | clusterin |
Gene Aliases: |
AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2 |
Location: |
Chr.8:27596917-27616717 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLU | NM_001831.3 | 7 | 1296 | NP_001822.3 |
| NR_038335.1 | 7 | 1431 | ||
| NR_045494.1 | 7 | 1290 | ||
| XM_006716284.2 | 7 | 3202 | XP_006716347.1 | |
| AK093399.1 | 8 | 1046 | ||
| AK301324.1 | 7 | 900 | ||
| AK313870.1 | 7 | 1185 | ||
| AY513288.1 | 7 | 1175 | AAT08041.1 | |
| BC010514.1 | 7 | 1178 | AAH10514.1 | |
| BC019588.2 | 6 | 1146 | AAH19588.1 | |
| BX648414.1 | 6 | 1896 | CAI45990.1 | |
| J02908.1 | 5 | 1028 | AAA51765.1 | |
| M25915.1 | 7 | 1305 | AAA35692.1 | |
| M64722.1 | 7 | 1162 | AAB06508.1 | |
| M74816.1 | 5 | 932 | AAA60321.1 | |
| X14723.1 | 6 | 1157 | CAA32847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831275 | Chr.8:27470773 - 27624088 on Build GRCh38 | Loss |
|
| esv3572652 | Chr.8:27589383 - 27606101 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map