Assay Details
Target Gene Details
Entrez Gene ID: | 9744 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 |
Gene Aliases: |
CENTB1 |
Location: |
Chr.17:7336506-7354944 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAP1 | NM_014716.3 | NP_055531.1 | ||
| AA356620.1 | ||||
| BC018543.1 | AAH18543.1 | |||
| BT009788.1 | AAP88790.1 | |||
| D30758.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv509650 | Chr.17:7299079 - 7338714 on Build GRCh38 | Insertion |
|
| nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map