Assay Details
Target Gene Details
Entrez Gene ID: | 153129 |
Gene Name: | solute carrier family 38 member 9 |
Gene Aliases: |
URLC11 |
Location: |
Chr.5:55625845-55712344 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830304 | Chr.5:55492810 - 55655813 on Build GRCh38 | Gain+Loss |
|
| nsv830305 | Chr.5:55543776 - 55743734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map