Assay Details
Target Gene Details
Entrez Gene ID: | 9315 |
Gene Name: | neuronal regeneration related protein |
Gene Aliases: |
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17 |
Location: |
Chr.5:111729303-111976931 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NREP | NM_001142474.1 | NP_001135946.1 | ||
| NM_001142475.1 | NP_001135947.1 | |||
| NM_001142476.1 | NP_001135948.1 | |||
| NM_001142477.1 | NP_001135949.1 | |||
| NM_001142478.1 | NP_001135950.1 | |||
| NM_001142479.1 | NP_001135951.1 | |||
| NM_001142480.1 | NP_001135952.1 | |||
| NM_001142481.1 | NP_001135953.1 | |||
| NM_001142482.1 | NP_001135954.1 | |||
| NM_001142483.1 | NP_001135955.1 | |||
| NM_004772.2 | NP_004763.1 | |||
| XM_006714732.1 | XP_006714795.1 | |||
| XM_006714733.2 | XP_006714796.1 | |||
| AF119859.1 | AAF69613.1 | |||
| AK307346.1 | ||||
| BC011050.1 | AAH11050.1 | |||
| BC019068.2 | AAH19068.1 | |||
| BC072013.1 | AAH72013.1 | |||
| BC072443.1 | AAH72443.1 | |||
| BX649051.1 | ||||
| U30521.1 | AAA74903.1 |
Target Gene Details
Entrez Gene ID: | 100505678 |
Gene Name: | STARD4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:111512226-111739726 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD4-AS1 | NR_040093.1 | |||
| CR749489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599367 | Chr.5:111588583 - 112267016 on Build GRCh38 | Gain |
|
| esv3606277 | Chr.5:111674439 - 111747485 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map